3-Methoxy-4-pyridineboronic acid pinacol ester 98+%

Supplier: Ambeed
Warning

A642842-100MG A642842-1G
77891-280EA 28 USD
77891-280 77891-281
3-Methoxy-4-pyridineboronic acid pinacol ester 98+%
3-Methoxy-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₃
MDL Number: MFCD13182222
CAS Number: 1243312-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR