Temsirolimus ≥99%

Supplier: Ambeed
Warning

Synonyms: (1R,2R,4R)-4-((R)-2-((3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,27-Dihydroxy-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-1,5,11,28,29-pentaoxo-1,4,5,6,9,10,11,12,13,14,21,22,23,24,25,26,27,28,29,31,32,33,34,34a-tetracosahydro-3H-23,27-epoxypyrido[2,1-c][1]oxa[4]azacyclohentriacontin-3-yl)propyl)-2-methoxycyclohexyl 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoate

A642622-5MG A642622-10MG
77669-754EA 46 USD
77669-754 77669-752
Temsirolimus ≥99%
Temsirolimus

Precaución: for Research Use Only

Formula: C₅₆H₈₇NO₁₆
MW: 1030.29 g/mol
MDL Number: MFCD00934421
CAS Number: 162635-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR