1-Benzyl-1H-pyrrol-2(5H)-one ≥98%

Supplier: Ambeed
Warning

A642541-100MG A642541-1G A642541-250MG
77891-277EA 60 USD
77891-277 77891-278 77891-279
1-Benzyl-1H-pyrrol-2(5H)-one ≥98%
1-Benzyl-1H-pyrrol-2(5H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MDL Number: MFCD09997905
CAS Number: 64330-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR