1,7-Diazaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥98%

Supplier: Ambeed
Warning

A641849-1G A641849-250MG A641849-5G
77873-037EA 194 USD
77873-037 77873-038 77873-039
1,7-Diazaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester ≥98%
1,7-Diazaspiro[4.4]nonane-7-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD09266216
CAS Number: 646055-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR