2-[(Dimethylamino)methyl]aniline ≥97%

Supplier: Ambeed
Warning

A641415-1G A641415-5G
77891-275EA 45 USD
77891-275 77891-276
2-[(Dimethylamino)methyl]aniline ≥97%
2-[(Dimethylamino)methyl]aniline

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MDL Number: MFCD00036084
CAS Number: 1904-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR