2-Fluoro-3-methyl-5-pyridineboronic acid pinacol ester ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A641205-1G A641205-250MG A641205-5G
77684-521EA 126 USD
77684-521 77684-522 77886-526
2-Fluoro-3-methyl-5-pyridineboronic acid pinacol ester ≥98%, Ambeed.Inc
2-Fluoro-3-methyl-5-pyridineboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₂H₁₇BFNO₂
MDL Number: MFCD11855990
CAS Number: 1150561-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR