1-Acetylpiperidine-4-carbaldehyde ≥98%

Supplier: Ambeed
Warning

A641115-100MG A641115-250MG
77594-964EA 21.53 USD
77594-964 77594-966
1-Acetylpiperidine-4-carbaldehyde ≥98%
1-Acetylpiperidine-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MW: 155.19 g/mol
MDL Number: MFCD11582881
CAS Number: 155826-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR