2'-Deoxyadenosine-5'-monophosphate ≥95%

Supplier: Ambeed
Warning

Synonyms: dAMP, ((2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

A641103-100MG A641103-1G A641103-250MG A641103-5G
77673-738EA 21 USD
77673-738 77673-740 77673-742 77673-744
2'-Deoxyadenosine-5'-monophosphate ≥95%
2'-Deoxyadenosine-5'-monophosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₅O₆P
MW: 331.22 g/mol
MDL Number: MFCD00005753
CAS Number: 653-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR