N-((S)-10-Benzyl-1-(((1S,9S)-9-ethyl-5-fluoro-9-hydroxy-4-methyl-10,13-dioxo-2,3,9,10,13,15-hexahydro-1H,12H-benzo[de]pyrano[3',4':6,7]indolizino[1,2-b]quinolin-1-yl)amino)-1,6,9,12,15-pentaoxo-3-oxa-5,8,11,14-tetraazahexadecan-16-yl)-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamide ≥98%

Supplier: Ambeed
Danger

A640944-10MG A640944-100MG A640944-50MG
77617-708EA 475.86 USD
77617-708 77749-315 77749-316
N-((S)-10-Benzyl-1-(((1S,9S)-9-ethyl-5-fluoro-9-hydroxy-4-methyl-10,13-dioxo-2,3,9,10,13,15-hexahydro-1H,12H-benzo[de]pyrano[3',4':6,7]indolizino[1,2-b]quinolin-1-yl)amino)-1,6,9,12,15-pentaoxo-3-oxa-5,8,11,14-tetraazahexadecan-16-yl)-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamide ≥98%
N-((S)-10-Benzyl-1-(((1S,9S)-9-ethyl-5-fluoro-9-hydroxy-4-methyl-10,13-dioxo-2,3,9,10,13,15-hexahydro-1H,12H-benzo[de]pyrano[3',4':6,7]indolizino[1,2-b]quinolin-1-yl)amino)-1,6,9,12,15-pentaoxo-3-oxa-5,8,11,14-tetraazahexadecan-16-yl)-6-(2,5-dioxo-2,5-di

Precaución: for Research Use Only

Formula: C₅₂H₅₆FN₉O₁₃
MW: 1034.05 g/mol
MDL Number: MFCD31657445
CAS Number: 1599440-13-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR