((2R,3R,4S,5R)-3-(Benzoyloxy)-5-(6-chloro-9H-purin-9-yl)-4-fluorotetrahydrofuran-2-yl)methyl benzoate ≥97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-9-(3,5-di-O-benzoyl-2-deoxy-2-fluoro-beta-D-arabinofuranosyl)-9H-purine

A640558-100MG A640558-1G A640558-250MG
77686-587EA 63 USD
77686-587 77686-588 77686-589
((2R,3R,4S,5R)-3-(Benzoyloxy)-5-(6-chloro-9H-purin-9-yl)-4-fluorotetrahydrofuran-2-yl)methyl benzoate ≥97%
((2R,3R,4S,5R)-3-(Benzoyloxy)-5-(6-chloro-9H-purin-9-yl)-4-fluorotetrahydrofuran-2-yl)methyl benzoate

Precaución: for Research Use Only

Formula: C₂₄H₁₈ClFN₄O₅
MW: 496.87 g/mol
MDL Number: MFCD15145215
CAS Number: 135473-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR