1,4-Bis(phenylethynyl)benzene ≥97%

Supplier: Ambeed
Warning

A640512-5G A640512-25G A640512-1G
77686-586EA 160 USD
77686-586 77901-068 77901-912
1,4-Bis(phenylethynyl)benzene ≥97%
1,4-Bis(phenylethynyl)benzene

Precaución: for Research Use Only

Formula: C₂₂H₁₄
MDL Number: MFCD00160829
CAS Number: 1849-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR