2-((6R)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

A640482-250MG A640482-50MG
77573-068EA 328.57 USD
77573-068 77617-706
2-((6R)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetic acid ≥98%
2-((6R)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇ClN₄O₂S
MW: 400.8819 g/mol
MDL Number: MFCD28167871
CAS Number: 202592-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR