Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-(tert-butyl)-4-hydroxyquinoline-3-carboxylate

A640331-1G A640331-250MG
77749-312EA 29 USD
77749-312 77749-313
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate ≥98%
Ethyl 6-tert-butyl-4-hydroxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₃
MW: 273.327 g/mol
MDL Number: MFCD24843129
CAS Number: 228728-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR