tert-Butyl 7-oxo-4-azaspiro[2.5]octane-4-carboxylate ≥95%

Supplier: Ambeed
Warning

A640058-1G A640058-100MG
77749-311EA 442 USD
77749-311 77749-310
tert-Butyl 7-oxo-4-azaspiro[2.5]octane-4-carboxylate ≥95%
tert-Butyl 7-oxo-4-azaspiro[2.5]octane-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD29763422
CAS Number: 1892578-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR