3-Indoxyl sulfate potassium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: Potassium 1H-indol-3-yl sulphate, 3-Indolyl sulphate potassium salt

A639949-1G A639949-250MG
77617-702EA 159.65 USD
77617-702 77617-704
3-Indoxyl sulfate potassium salt ≥98%
3-Indoxyl sulfate potassium salt

Precaución: for Research Use Only

Formula: C₈H₆KNO₄S
MW: 251.31 g/mol
MDL Number: MFCD00037931
CAS Number: 2642-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR