7-Fluorobenzo[d]thiazole-2(3H)-thione ≥95%

Supplier: Ambeed
Warning

A639816-1G A639816-250MG A639816-5G
77617-700EA 20.6 USD
77617-700 77749-308 77749-309
7-Fluorobenzo[d]thiazole-2(3H)-thione ≥95%
7-Fluorobenzo[d]thiazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₇H₄FNS₂
MW: 185.24 g/mol
MDL Number: MFCD08692055
CAS Number: 154327-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR