N,N-Dimethyl-1-dodecylamine N-oxide ≥98%

Supplier: Ambeed
Danger

A639629-1G A639629-25G A639629-5G A639629-100G
77602-626EA 23.58 USD
77602-626 77602-628 77602-630 77669-734
N,N-Dimethyl-1-dodecylamine N-oxide ≥98%
N,N-Dimethyl-1-dodecylamine N-oxide

Precaución: for Research Use Only

Formula: C₁₄H₃₁NO
MDL Number: MFCD00002049
CAS Number: 1643-20-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR