tert-Butyl (32-amino-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontyl)carbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG10-CH2CH2NH2

A639524-100MG A639524-250MG
77598-642EA 134.28 USD
77598-642 77783-564
tert-Butyl (32-amino-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontyl)carbamate ≥95%
tert-Butyl (32-amino-3,6,9,12,15,18,21,24,27,30-decaoxadotriacontyl)carbamate

Precaución: for Research Use Only

Formula: C₂₇H₅₆N₂O₁₂
MDL Number: MFCD31656920
CAS Number: 1347704-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR