3-Benzyloxy-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-(Benzyloxy)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Benzyloxy)-4-fluorophenylboronic acid pinacolester, 3-Benzyloxy-4-fluorobenzeneboronic acid pinacol ester

A639348-100MG A639348-1G A639348-250MG A639348-5G
77783-560EA 123 USD
77783-560 77783-561 77783-562 77783-563
3-Benzyloxy-4-fluorophenylboronic acid pinacol ester ≥97%
3-Benzyloxy-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BFO₃
MDL Number: MFCD23098800
CAS Number: 2121515-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR