tert-Butyl 3,3-bis(hydroxymethyl)azetidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A639220-100MG A639220-1G A639220-250MG A639220-5G
77749-303EA 22 USD
77749-303 77749-304 77749-305 77749-306
tert-Butyl 3,3-bis(hydroxymethyl)azetidine-1-carboxylate ≥98%
tert-Butyl 3,3-bis(hydroxymethyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
MDL Number: MFCD24470652
CAS Number: 1016233-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR