6-Chloro-1-methyl-2-indoleboronic acid pinacol ester ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
A638928-1G
A638928-250MG
A638928-5G
77886-521EA
211
USD
77886-521
77886-522
77886-523
6-Chloro-1-methyl-2-indoleboronic acid pinacol ester ≥98%
6-Chloro-1-methyl-2-indoleboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₁₉BClNO₂ |
MDL Number:
MFCD11504964 CAS Number: 1073353-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...