2'-Deoxycytidine-5'-monophosphate disodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium ((2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl phosphate, Disodium 2'-deoxy-5'-O-phosphonatocytidine

A637172-100MG A637172-1G A637172-250MG
77627-270EA 18.54 USD
77627-270 77627-272 77627-274
2'-Deoxycytidine-5'-monophosphate disodium salt ≥98%
2'-Deoxycytidine-5'-monophosphate disodium salt

Precaución: for Research Use Only

Formula: C₉H₁₂N₃Na₂O₇P
MDL Number: MFCD00069774
CAS Number: 13085-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR