2-(4-Ethylphenoxy)-N-(4-fluoro-3-nitrophenyl)acetamide ≥97%

Supplier: Ambeed
Warning

A637118-100MG A637118-250MG A637118-50MG
77783-554EA 268 USD
77783-554 77783-555 77783-556
2-(4-Ethylphenoxy)-N-(4-fluoro-3-nitrophenyl)acetamide ≥97%
2-(4-Ethylphenoxy)-N-(4-fluoro-3-nitrophenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₆H₁₅FN₂O₄
MDL Number: MFCD02219567
CAS Number: 425613-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR