2-((R)-4-((3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulfonic acid ≥98%

Supplier: Ambeed
Warning

A637009-10MG A637009-5MG
77613-976EA 19.57 USD
77613-976 77613-978
2-((R)-4-((3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulfonic acid ≥98%
2-((R)-4-((3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethanesulfonic acid

Precaución: for Research Use Only

Formula: C₂₆H₄₅NO₆S
MDL Number: MFCD00232922
CAS Number: 516-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR