Fmoc-L-homopropargylglycine ≥98%

Supplier: Ambeed
Warning

Synonyms: N,Fmoc-L-homopropargylglycine, Fmoc-HoGly(Propargyl)-OH, (2S)-2-(Fmoc-amino)-5-hexynoic acid

A636800-250MG A636800-100MG A636800-1G
77622-410EA 299.73 USD
77622-410 77783-552 77783-553
Fmoc-L-homopropargylglycine ≥98%
Fmoc-L-homopropargylglycine

Precaución: for Research Use Only

Formula: C₂₁H₁₉NO₄
MW: 349.38 g/mol
MDL Number: MFCD11052899
CAS Number: 942518-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR