SGC-CBP30 ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-4-(1-(2-(3-Chloro-4-methoxyphenethyl)-5-(3,5-dimethylisoxazol-4-yl)-1H-benzo[d]imidazol-1-yl)propan-2-yl)morpholine

A636668-1MG A636668-2MG
77613-972EA 51.5 USD
77613-972 77613-974
SGC-CBP30 ≥98%
SGC-CBP30

Precaución: for Research Use Only

Formula: C₂₈H₃₃ClN₄O₃
MW: 509.04 g/mol
MDL Number: MFCD26792586
CAS Number: 1613695-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR