Amino-PEG12-Boc ≥95%

Supplier: Ambeed
Warning

A635623-100MG A635623-50MG A635623-1G A635623-250MG
77598-640EA 65.6 USD
77598-640 77622-408 77783-548 77783-549
Amino-PEG12-Boc ≥95%
Amino-PEG12-Boc

Precaución: for Research Use Only

Formula: C₃₁H₆₃NO₁₄
MDL Number: MFCD11041147
CAS Number: 1383814-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR