3-Bromo-3-methyl-2-butanone ≥98%

Supplier: Ambeed

A635555-25G A635555-5G
77882-474EA 334 USD
77882-474 77882-475
3-Bromo-3-methyl-2-butanone ≥98%
3-Bromo-3-methyl-2-butanone

Precaución: for Research Use Only

Formula: C₅H₉BrO
MDL Number: MFCD06661470
CAS Number: 2648-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR