tert-Butyl (38-amino-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaoctatriacontyl)carbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG12-NH2

A635248-250MG A635248-100MG A635248-1G A635248-5G
77622-406EA 199.82 USD
77622-406 77783-545 77783-546 77783-547
tert-Butyl (38-amino-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaoctatriacontyl)carbamate ≥95%
tert-Butyl (38-amino-3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaoctatriacontyl)carbamate

Precaución: for Research Use Only

Formula: C₃₁H₆₄N₂O₁₄
MDL Number: MFCD31656921
CAS Number: 1642551-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR