3-Bromo-5-methyl-1H-1,2,4-triazole ≥98%

Supplier: Ambeed
Warning

A635026-1G A635026-5G A635026-100MG
77749-299EA 308 USD
77749-299 77749-300 77749-298
3-Bromo-5-methyl-1H-1,2,4-triazole ≥98%
3-Bromo-5-methyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₃H₄BrN₃
MDL Number: MFCD13188583
CAS Number: 26557-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR