3,4-Difluoro-5-methoxyphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (3,4-Difluoro-5-methoxyphenyl)boronic acid

A635022-100MG A635022-1G A635022-250MG
77873-030EA 29 USD
77873-030 77873-031 77873-032
3,4-Difluoro-5-methoxyphenylboronic acid ≥98%
3,4-Difluoro-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MDL Number: MFCD09907960
CAS Number: 925910-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR