Methylene blue hydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Methylthioninium chloride hydrate, Basic Blue 9, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate, C.I. 52015, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride xhydrate

A634813-100G A634813-25G
77613-960EA 33.99 USD
77613-960 77613-962
Methylene blue hydrate ≥98%
Methylene blue hydrate

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClN₃S·xH₂O
MDL Number: MFCD00150006
CAS Number: 122965-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR