(R)-5,7-Dihydroxy-3-(4-methoxybenzyl)-6,8-dimethylchroman-4-one ≥98%
Supplier: Ambeed
|
Warning
|
A634435-10MG
A634435-1MG
A634435-5MG
77893-271EA
231
USD
77893-271
77893-272
77893-273
(R)-5,7-Dihydroxy-3-(4-methoxybenzyl)-6,8-dimethylchroman-4-one ≥98%
(R)-5,7-Dihydroxy-3-(4-methoxybenzyl)-6,8-dimethylchroman-4-one
Precaución: for Research Use Only
Formula:
C₁₉H₂₀O₅ |
MDL Number:
MFCD20487835 CAS Number: 74805-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...