2-Cyclopropoxyacetic acid ≥95%

Supplier: Ambeed
Danger

A634188-100MG A634188-1G A634188-250MG
77891-267EA 292 USD
77891-267 77891-268 77891-269
2-Cyclopropoxyacetic acid ≥95%
2-Cyclopropoxyacetic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
MDL Number: MFCD19231341
CAS Number: 246869-07-8
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR