(S)-a-[Fmoc-(methyl)amino]cyclohexaneacetic acid ≥95%

Supplier: Ambeed
Warning

A633844-1G A633844-250MG A633844-100MG
77749-294EA 323 USD
77749-294 77749-295 77749-293
(S)-a-[Fmoc-(methyl)amino]cyclohexaneacetic acid ≥95%
(S)-a-[Fmoc-(methyl)amino]cyclohexaneacetic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₄
MDL Number: MFCD31704916
CAS Number: 925240-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR