(R)-10-Methyl-3-(6-methylpyridin-3-yl)-9,10,11,12-tetrahydro-8H-[1,4]diazepino[5',6':4,5]thieno[3,2-f]quinolin-8-one ≥99%

Supplier: Ambeed
Warning

Synonyms: PF-3644022

A633788-50MG A633788-100MG A633788-250MG
77573-058EA 337 USD
77573-058 77686-581 77901-653
(R)-10-Methyl-3-(6-methylpyridin-3-yl)-9,10,11,12-tetrahydro-8H-[1,4]diazepino[5',6':4,5]thieno[3,2-f]quinolin-8-one ≥99%
(R)-10-Methyl-3-(6-methylpyridin-3-yl)-9,10,11,12-tetrahydro-8H-[1,4]diazepino[5',6':4,5]thieno[3,2-f]quinolin-8-one

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₄OS
MW: 374.45 g/mol
MDL Number: MFCD21363374
CAS Number: 1276121-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR