t-Boc-N-Amido-PEG12-acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG12-CH2CH2COOH, 2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26,29,32,35,38,41-tridecaoxa-5-azatetratetracontan-44-oic acid

A633699-50MG A633699-100MG
77622-402EA 101.97 USD
77622-402 77783-539
t-Boc-N-Amido-PEG12-acid ≥95%
t-Boc-N-Amido-PEG12-acid

Precaución: for Research Use Only

Formula: C₃₂H₆₃NO₁₆
MDL Number: MFCD13185149
CAS Number: 1415981-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR