4-((6-(Acryloyloxy)hexyl)oxy)phenyl 4-methoxybenzoate ≥97%
Supplier: Ambeed
|
Warning
|
A633692-1G
A633692-5G
77747-303EA
139
USD
77747-303
77747-304
4-((6-(Acryloyloxy)hexyl)oxy)phenyl 4-methoxybenzoate ≥97%
4-((6-(Acryloyloxy)hexyl)oxy)phenyl 4-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₂₃H₂₆O₆ MW: 398.44 g/mol |
CAS Number:
130953-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...