Lactose octaacetate ≥95%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)-5-(((2S,3R,4S,5S,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triyl triacetate

A633379-25G A633379-1G
77573-054EA 198.79 USD
77573-054 77669-716
Lactose octaacetate ≥95%
Lactose octaacetate

Precaución: for Research Use Only

Formula: C₂₈H₃₈O₁₉
MW: 678.593 g/mol
MDL Number: MFCD00064040
CAS Number: 6291-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR