Fmoc-4-trifluoromethyl-L-homophenylalanine ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-HoPhe(4-CF₃)-OH

A632229-100MG A632229-50MG
77783-531EA 66 USD
77783-531 77783-532
Fmoc-4-trifluoromethyl-L-homophenylalanine ≥97%
Fmoc-4-trifluoromethyl-L-homophenylalanine

Precaución: for Research Use Only

Formula: C₂₆H₂₂F₃NO₄
MDL Number: MFCD07372279
CAS Number: 1260591-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR