2-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-5-one ≥98%

Supplier: Ambeed
Warning

A632146-250MG A632146-1G
77573-050EA 115.36 USD
77573-050 77594-944
2-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-5-one ≥98%
2-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-5-one

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.59 g/mol
MDL Number: MFCD11557189
CAS Number: 1092301-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR