Methyl-4-iodobutanoate ≥97% stabilised

Supplier: Ambeed
Warning

Synonyms: Methyl 4-iodobutanoate, Methyl 4-iodobutyrate

Stabilization: Stabilized with Copper chip

A632095-5G A632095-1G
77686-577EA 345 USD
77686-577 77900-922
Methyl-4-iodobutanoate ≥97% stabilised
Methyl-4-iodobutanoate

Precaución: for Research Use Only

Formula: C₅H₉IO₂
MDL Number: MFCD00674066
CAS Number: 14273-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR