(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)-2,6-dimethylphenyl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A632075-100MG A632075-1G A632075-250MG A632075-50MG
77891-255EA 210 USD
77891-255 77891-256 77891-257 77891-258
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)-2,6-dimethylphenyl)propanoic acid ≥95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)-2,6-dimethylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₃₀H₃₃NO₅
MDL Number: MFCD20133793
CAS Number: 1043043-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR