1-Fluoropyridiniumtrifluoromethanesulfonate ≥98%

Supplier: Ambeed
Danger

Synonyms: N-Fluoropyridinium triflate, 1-Fluoropyridiniumtriflate, N-Fluoropyridinium trifluoromethanesulphonate

A632020-25G A632020-1G
77573-046EA 156 USD
77573-046 77594-942
1-Fluoropyridiniumtrifluoromethanesulfonate ≥98%
1-Fluoropyridinium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₆H₅F₄NO₃S
MW: 247.17 g/mol
MDL Number: MFCD00013458
CAS Number: 107263-95-6
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR