Platinum(IV) iodide ≥98%

Supplier: Ambeed
Danger

Synonyms: Platinum tetraiodide, Platinum(iv)iodide

A631150-250MG A631150-100MG
77669-710EA 42 USD
77669-710 77880-339
Platinum(IV) iodide ≥98%
Platinum(IV) iodide

Precaución: for Research Use Only

Formula: I₄Pt
MW: 702.7
Density: 6.06
MDL Number: MFCD00016292
CAS Number: 7790-46-7
UN: 2923
ADR: 8,6.1 Ⅱ
Merck Index: 12,07685

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR