(2-Chloro-4-phenoxyphenyl)(4-(((3R,6S)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)amino)-3H-pyrrolo[2,3-d]pyrimidin-5-yl)methanone ≥99%

Supplier: Ambeed
Warning

Synonyms: ARQ 531

A630399-10MG A630399-1MG A630399-5MG
77598-622EA 85.08 USD
77598-622 77598-624 77598-626
(2-Chloro-4-phenoxyphenyl)(4-(((3R,6S)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)amino)-3H-pyrrolo[2,3-d]pyrimidin-5-yl)methanone ≥99%
(2-Chloro-4-phenoxyphenyl)(4-(((3R,6S)-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)amino)-3H-pyrrolo[2,3-d]pyrimidin-5-yl)methanone

Precaución: for Research Use Only

Formula: C₂₅H₂₃ClN₄O₄
MW: 478.93 g/mol
MDL Number: MFCD31692379
CAS Number: 2095393-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR