6-(Trifluoromethyl)-4-quinolinol ≥98%

Supplier: Ambeed
Warning

A630330-5G A630330-25G
77602-592EA 23.58 USD
77602-592 77880-338
6-(Trifluoromethyl)-4-quinolinol ≥98%
6-(Trifluoromethyl)-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
MDL Number: MFCD00209673
CAS Number: 49713-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR