2-Azabicyclo[2.2.1]heptane hydrochloride ≥98%

Supplier: Ambeed
Warning

A629753-1G A629753-250MG
77891-246EA 97 USD
77891-246 77891-247
2-Azabicyclo[2.2.1]heptane hydrochloride ≥98%
2-Azabicyclo[2.2.1]heptane hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClN
MDL Number: MFCD13516979
CAS Number: 63838-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR