2-Methyl-[1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A629720-100MG A629720-250MG
77749-286EA 13 USD
77749-286 77749-287
2-Methyl-[1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid ≥97%
2-Methyl-[1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.1601 g/mol
MDL Number: MFCD22092796
CAS Number: 1538440-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR