Benzo[b]thiophene-3-acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (Benzo[b]thiophen-3-yl)acetic acid

A629228-250MG A629228-5G A629228-1G
77669-706EA 18 USD
77669-706 77669-708 77880-335
Benzo[b]thiophene-3-acetic acid ≥98%
Benzo[b]thiophene-3-acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MDL Number: MFCD00051637
CAS Number: 1131-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR